Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-7-nitrobenzofurazan

Supplier: AAT BIOQUEST INC
Warning
Synonyms: NBD-F4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole

821
76481-024EA 118.54 USD InStock
76481-024
4-Fluoro-7-nitrobenzofurazan
4-Fluoro-7-nitrobenzofurazan
NBD-F has properties and applications similar to those of NBD-Cl.


Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
MDL Number: MFCD00010196
CAS Number: 29270-56-2
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty