Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-7-nitrobenzofurazan

Supplier: AAT BIOQUEST INC
Warning
Synonyms: NBD-ClNBD chloride4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole

825
76481-026EA 93.54 USD InStock
76481-026
4-Chloro-7-nitrobenzofurazan
4-Chloro-7-nitrobenzofurazan
NBD-Cl is widely used to label peptides, proteins, drugs and other biomolecules.


Formula: C₆H₂ClN₃O₃
MW: 199.55 g/mol
MDL Number: MFCD00005808
CAS Number: 10199-89-0
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty