Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biotin hydrazide ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide

3007
76481-616EA 93.54 USD InStock
76481-616
Biotin hydrazide ≥95%
Biotin hydrazide
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.


Formula: C₁₀H₁₈N₄O₂S
MW: 258.34 g/mol
MDL Number: MFCD00078532
CAS Number: 66640-86-6
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty