Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DSG (Disuccinimidyl glutarate) ≥90%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) glutarateDi(N-succinimidyl) glutarateDSG

4520
76481-698EA 93.54 USD InStock
76481-698
DSG (Disuccinimidyl glutarate) ≥90%
DSG (Disuccinimidyl glutarate)
DSG is disuccinimidyl glutarate, a simple homobifunctional crosslinker based on the amine-reactive N-hydroxysuccinimide (NHS) ester group at both ends of a 5-atom (7.7 angstrom) spacer arm.


Formula: C₁₃H₁₄N₂O₈
MW: 326.26 g/mol
MDL Number: MFCD00153597
CAS Number: 79642-50-5
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty