Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stains-all ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

10050
76481-844EA 93.54 USD InStock
76481-844
Stains-all ≥95%
Stains-all
Stains-all is suitable for differential staining of nucleic acids and proteins.


Formula: C₃₀H₂₇BrN₂S₂
MW: 559.584 g/mol
MDL Number: MFCD00078788
CAS Number: 7423-31-6
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty