Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ac-Leu-Glu-His-Asp-AMC ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

13426
76482-096EA 180.84 USD InStock
76482-096
Ac-Leu-Glu-His-Asp-AMC ≥95%
Ac-Leu-Glu-His-Asp-AMC
Caspase 9 is a member of the CED-3 subfamily of the caspase family of cysteine proteases that play an essential role in the execution phase of apoptosis.


Formula: C₃₃H₄₁N₇O₁₁
MDL Number: MFCD01862664
CAS Number: 292633-16-0
ADR: 5,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty