Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl-β-D-glucopyranoside ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: 4-Methylumbelliferyl beta-D-GlucopyranosideMUGlucosid4-MU-β-D-Glc

14014
76482-296EA 93.54 USD InStock
76482-296
4-Methylumbelliferyl-β-D-glucopyranoside ≥95%
4-Methylumbelliferyl-beta-D-glucopyranoside
4-Methylumbelliferyl-beta-D-glucoside is a fluorogenic MU substrate for detecting glucosidase in cell extracts and purified enzyme preparations using a fluorescence microplate reader or fluorometer.


Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
MDL Number: MFCD00063694
CAS Number: 18997-57-4
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty