Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Ethoxyresorufin ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: EthoxyresorufinResorufin ethyl ether

15023
76482-316EA 93.54 USD InStock
76482-316
7-Ethoxyresorufin ≥95%
7-Ethoxyresorufin
This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.


Formula: C₁₄H₁₁NO₃
MW: 241.246 g/mol
MDL Number: MFCD00037661
CAS Number: 5725-91-7
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty