Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Diaminonaphthalene ≥95%

Supplier: AAT BIOQUEST INC
Warning
Synonyms: DAN2,3-Naphthalenediamine2,3-naphthylenediamine

15221
76482-334EA 93.54 USD InStock
76482-334
2,3-Diaminonaphthalene ≥95%
2,3-Diaminonaphthalene
The role of nitric oxide (NO) as a putative mediator of neuronal death can be understood best if NO is detected directly.


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
MDL Number: MFCD00004116
CAS Number: 771-97-1
ADR: 5,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty