Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester

Supplier: AAT BIOQUEST INC
Synonyms: N-Succinimidyl 7-Hydroxy-4-methylcoumarin-3-acetate7-Hydroxy-4-methylcoumarin-3-acetic acid N-Succinimidyl ester4-Methylumbelliferone-3-acetic acid N-Succinimidyl esterN-Succinimidyl 4-Methylumbelliferone-3-acetate

556
76480-978EA 118.54 USD InStock
76480-978
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid, SE is an amine-reactive blue fluorophore that has pH-dependent and environment-sensitive fluorescence.

  • Multiple Bioconjugation Applications
  • pH-independent Fluorescence

Caution: For Research Use Only


Formula: C₁₆H₁₃NO₇
MW: 331.28 g/mol
MDL Number: MFCD00467590
CAS Number: 96735-88-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty