Would you like to log in again?
Whatman™ Grade 520 bII Filter Papers for Technical Use, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10331687 10331687
10035-638PK 1308.3 USD InStock
10035-638 CA11013-784
Whatman™ Grade 520 bII Filter Papers for Technical Use, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A thick paper with high wet strength offering a very high flow rate.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Also available prepleated as Grade 520 bII ½.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty