Would you like to log in again?
Whatman™ Grade 0905 Filter Papers for Technical Use, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10334987 10334987
10035-788PK 706.37 USD InStock
10035-788 CA11013-260
Whatman™ Grade 0905 Filter Papers for Technical Use, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A creped paper for coarse particles, offers a very high filtration speed.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty