Would you like to log in again?
Whatman™ Grade 2294 Filter Papers for Technical Use, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10342860 10342810 10342860 10342810
10034-766PK 94.96 USD InStock
CA11013-412 10034-766 CA11013-272 10034-764
Whatman™ Grade 2294 Filter Papers for Technical Use, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A very thick filter card with high wet strength. Offers very high flow rate and retains medium to coarse particles.
  • With central hole
The applications range from the filtration of beverages to the purification of electroplating baths.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty