Would you like to log in again?
Whatman™ Grade 2589 D Filter Papers for Technical Use, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10343978 10343976 10343978 10343976
76423-766PK 318.61 USD InStock
CA11013-274 CA76423-766 76423-766 10034-776
Whatman™ Grade 2589 D Filter Papers for Technical Use, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A very thick filter with high wet strength. Offers medium to slow flow rate and retains very fine precipitates.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty