Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Multi Stirrus™ Magnetic Stirrer, 230 V

Supplier: V&P Scientific
VP710D3
MSPP-VP710D3EA 2641.35 USD InStock
MSPP-VP710D3
Multi Stirrus™ Magnetic Stirrer, 230 V
Stirrers Magnetic Stirrers
The Multi stirrus systems can be used as vertical tumble stirrers, lateral vortex tumble stirrers and conventional magnetic stirrers.

  • 230 V model
  • Can stir in any vessel regardless of the size or shape
  • Can stir in syringes to keep bar coded beads in suspension for drop seq assays
  • Complete stirring of large numbers of samples
  • Can stir chemical reactions to speed completion
  • Can run multiple experiments in parallel
  • Simple to operate, full speed control
  • Can set exact RPM for reproducible results day to day
  • Designed to be compatible with most robotic systems
  • Flexibility of stir deck alignment

A vertical tumble stirrer causes stir elements of all shapes and sizes to tumble end over end inside each of several vessels. A lateral vortex stirrer causes stir elements of all shapes and sizes to spin laterally within each of many vessels. A conventional magnetic stirrer causes one stir bar to spin horizontally in the center of a single bottle or vessel.

This system can be used as vertical tumble stirrer to mix samples in several, micro centrifuge tubes, test tubes, vials, syringes and to keep particulates in suspension in syringes troughs and reservoirs. Can be used as a lateral vortex tumble stirrer to mix many samples in micro centrifuge tubes, test tubes, vials and syringes and can also be used as a conventional horizontal stirrer to mix a single bottle or vessel.

This system can stir even 100000 centistoke viscous solutions (6.6 times more viscous than honey) and even thick slurries and emulsions.

Magnetic cylinder length: 1".

Caution: 230 V model.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty