Would you like to log in again?
SP Bel-Art Filter Paper Discs for Buchner Table-Top Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H14632-0018 H14632-0024 H14632-0010 H14632-0036
16126-590PK 1523.04 USD InStock
16126-590 30309-000 30309-002 30309-004
SP Bel-Art Filter Paper Discs for Buchner Table-Top Funnels, Bel-Art Products, a part of SP
Filters Filter Paper Quantitative Filter Paper
These heavy bond grade filter paper discs are custom-sized to fit table-top Buchner funnels and have a thickness of 0.30 mm.
  • Made from creped white cellulose to improve porosity
  • Flow rate: 90 ml per minute
  • Wet burst strength is greater than 68 KPa; able to retain particles as small as 3 microns
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty