Would you like to log in again?
VSV-Pseudovirus_MERS-CoV Luciferase, ReVacc Scientific
Supplier: ReVacc Scientific
T53L53-1ML T53L53-200UL
76515-506EA 2225.77 USD InStock
76515-506 76515-504
VSV-Pseudovirus_MERS-CoV Luciferase, ReVacc Scientific
Vectors, Plasmids and Libraries
This pseudotyped virus uses recombinant Vesicular Stomatitis Virus (rVSV) to carry S protein of MERS-CoV (GenBank: JX869059.2), with 16-aa cytoplasmic tail truncation for optimal infection.
Infection of cells with this pseudotyped virus carrying luciferase reporter results in high level luciferase activity. See the titration result showing the undiluted pseudovirus generates signal ~100000-fold higher than uninfected control (cell alone as background). The neutralizing activity of a control mAb and one published mAb called MERS27 was tested by using this pseudotyped virus.
As pseudovirus infectivity of rVSV without its original G is restricted to a single round of replication, the pseudotypes can be handled using BSL-2 containment practices. Applications include Anti-SARS-CoV-2 neutralizing antibody screening at high throughput, Anti-SARS-CoV-2 drug screening at high throughput and SARS-CoV-2 pseudovirus transduction of target cells for viral entry and functional studies.
Packaging: Shipping with dry ice. Require −80 °C storage. Multiple freeze/Thaw cycles will reduce its sensitivity. Recommend only one cycle. Aliquot after the first thaw.
Caution: Handle it in biosafety cabinet in BSL-2. Contacted tips and tubes should be decontaminated by 10% disinfecting bleach. For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty