Would you like to log in again?
Metacarb columns, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
A5200 A5200
AGA5200EA 2596 USD InStock
AGA5200 CAAGA5200
Metacarb columns, Agilent Technologies
Chromatography Columns
The Agilent MetaCarb line of carbohydrate columns provides high resolution columns for applications requiring high performance Carbohydrate and Organic Acid Analysis.
  • MetaCarb columns provide a wide range of selectivities for carbohydrate and organic analysis
  • Column selectivities provide superior reproducibility and durability
  • The columns are very rugged, operate at low backpressure, and provide outstanding column lifetime
MetaCarb columns contain sulfonated polystyrene resins in the Calcium (87C), Hydrogen (87H), and Lead (87P) forms, and provide a wide range of selectivities for carbohydrate and organic analysis. Column selectivities provide superior reproducibility and durability. The MetaCarb 87 series is based on an 8 to 10 μl, 8% crosslinked styrene divinylbenzene (SDVB) resin. The columns are very rugged, operate at low backpressure, and provide outstanding column lifetime.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty