Would you like to log in again?
PLgel Olexis, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
PL1110-1400 PL1110-1400
AGPL1110-1400EA 691 USD InStock
AGPL1110-1400 CAAGPL11101400
PLgel Olexis, Agilent Technologies
Chromatography Columns
PLgel Olexis GPC columns are designed for analysis of high-mol wt polymers and high-temperature GPC.
  • Optimized design for analysis of polyolefins, polyethylene, polypropylene, and high-performance polymers
  • Offers efficiency, long lifetime, and resolution without the risk of degrading samples
  • Compatible with 1,2,4-trichlorobenzene (TCB), dichlorobenzene (DCB), chloronaphthalene, and green solvents such as butylal
PLgel Olexis GPC columns are optimized for the analysis of high-molecular weight polymers and high-temperature GPC analysis. Olexis columns are the industry standard for analysis of polyolefin, polyethylene, polypropylene, and high-performance polymers. Even at elevated temperatures, the PL Olexis offers efficiency, long lifetime, and resolution without the risk of shear degradation.
Columns are compatible with 1,2,4-trichlorobenzene (TCB), dichlorobenzene (DCB), chloronaphthalene, and green solvents such as butylal.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty