Would you like to log in again?
CrossLab CP-Chirasil Val Columns, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
CP7495 CP7495
AGCP7495EA 1518 USD InStock
AGCP7495 CAAGCP7495
CrossLab CP-Chirasil Val Columns, Agilent Technologies
Chromatography Columns
Designed for the separation of optically active compounds, especially amino acids.
  • Designed and tested for separations of optically active compounds, including amino acids
  • Both antipode phases available (D and L) for maximum versatility
  • Stabilized phase, over 50% crosslinked for great longevity
  • Low bleed
  • On Chirasil-L Val, D-amino acids elute first, while on Chirasil-D-Val, this elution order is reversed - especially valuable for optical purity determination
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Chirasil Val columns are designed for the separation of optically active compounds, especially amino acids. They have lower bleed levels than other phases, with a Tmax of 200 °C, isothermally and programmed. Both antipodes of the phase are available. On Chirasil-L-Val, D-amino acids elute before the L-amino acids, while on Chirasil-D-Val this elution order is reversed. This is especially valuable when determining the optical purity of compounds. Selecting the column from which the minor component elutes before the major enantiomer results in the lowest detection levels.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty