Would you like to log in again?
DB-35ms Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
128-3812 128-3812 125-3837 122-3862 122-3831 128-3822-INT 128-3822 122-3832 123-3812 122-3812 122-3832 122-3812 123-3812 125-3832 125-3837 122-3862 123-3832 128-3822-INT 128-3822 125-3832 123-3832 122-3831
AG123-3812EA 639.53 USD InStock
CAAG125-3837 CAAG122-3862 CAAG122-3831 AG123-3812 CA122-3812 AG128-3822-INT AG123-3832 AG122-3832 CA122-3832 AG128-3812 CA128-3812 AG128-3822 AG122-3831 AG122-3862 CA125-3832 AG125-3832 CAAG1283822INT AG125-3837 CAAG128-3822 AG122-3812 CA123-3832 CAAG123-3812
DB-35ms Columns, Agilent Technologies
Chromatography Columns
Virtually equivalent to a (35%-phenyl) methylpolysiloxane, with excellent inertness and low bleed, ideal for confirmation analysis.
  • Virtually equivalent to (35%-phenyl)-methylpolysiloxane
  • Mid polarity with very low bleed characteristics, ideal for GC/MS
  • Extended temperature limit of 340/360 °C
  • Excellent inertness for active compounds
  • Ideal for confirmation analyses
  • Close equivalent to USP phase G42
  • Bonded, crosslinked, and solvent rinsible
  • Replaces HP-35ms
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-35ms, a close equivalent to USP phase G42, is precision engineered to deliver excellent performance in a range of GC/MS applications, particularly in confirmation analysis. These bonded, crosslinked, mid-polarity columns operate over an extended temperature range up to 340/360 °C. They feature very low-bleed characteristics and excellent inertness and are particularly well suited for analysis of active compounds.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty