Would you like to log in again?
Select Mineral Oil Columns, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
CP7491 CP7491
AGCP7491EA 563 USD InStock
CAAGCP7491 AGCP7491
Select Mineral Oil Columns, Agilent Technologies
Chromatography Columns
Simple and reliable, with a stabilized nonpolar bonded phase specifically for fast mineral oil analysis.
  • Optimized selectivity for reliable total petroleum hydrocarbon (TPH) results per DIN H53 N-ISO 9377-2 methods
  • C4 to C40 hydrocarbons can be analyzed in less than ten minutes
  • Low bleed
  • Available in fused silica or UltiMetal
  • Fast run time
  • Available in economical 3 and 6 packs
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phase: Rtx-Mineral Oil
Agilent J&W Select Mineral Oil provides a simple and reliable option for total petroleum hydrocarbon analysis, a routine technique in many environmental laboratories, where there are often many samples needing to be screened. The column features a stabilized nonpolar bonded phase specifically for fast mineral oil analysis, is temperature stable up to 375/400 °C, and provides rapid analysis according to DIN H53 and DIN-EN-ISO 9377-2 methods.
Due to the temperature stability this column, C4–C40 hydrocarbons can be analyzed in less than ten minutes, and the high temperature stability permits faster bake-out. For optimal injection performance, use with the special 4-m retention gap (CP8015).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty