Would you like to log in again?
J&W VF-35ms Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP8884 CP8877 CP8874 CP8877 CP8884 CP8874
AGCP8884EA 857 USD InStock
AGCP8884 CAAGCP8877 CAAGCP8884 CAAGCP8874 AGCP8874 AGCP8877
J&W VF-35ms Columns, Agilent Technologies
Chromatography Columns
Has a medium polarity stationary phase and is ideal for trace environmental and chemical analyses, and as a confirmation column.
  • Stabilized arylene-modified equivalent of a 35% phenylmethyl phase
  • Ideal for dual-column confirmational analyses
  • Medium polarity ideal for trace environmental and chemical analyses
  • 0.15 mm Int. Ø columns available for high efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
The VF-35ms is a low-bleed, highly stable column with a programmable maximum temperature of 360 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty