Would you like to log in again?
Imapex™ Peroxide Cured Silicone Tubing, Foxx Life Sciences
Supplier: Foxx Life Sciences
PEX00790127025 PEX01000180025 PEX00600120050 PEX00790127050 PEX02500310050 PEX00950159025 PEX01000140100 PEX00900150100 PEX00300060050 PEX00950127025 PEX00500100050 PEX01270222050 PEX00800120100 PEX00400090050 PEX00800160100 PEX00950143050 PEX01200180050 PEX00950190025 PEX01000160100 PEX00950143025 PEX00500100100 PEX00400070025 PEX00950127050 PEX01500210050 PEX01270190050 PEX01590254050 PEX00800110025 PEX00300080025 PEX00900150050 PEX00500090100 PEX00300060100 PEX00600100100 PEX00500110025 PEX00950190050 PEX01000180050 PEX00300060025 PEX01200180025 PEX00500080050 PEX00900160025 PEX00320063025 PEX01600240050 PEX01900270050 PEX00400100050 PEX00400060025 PEX00630127100 PEX00800120050 PEX00480079025 PEX00300080100 PEX00300070100 PEX01270175025 PEX00300080050 PEX01200160025 PEX00900160100 PEX00320063100 PEX00900160050 PEX00480095100 PEX01270190100 PEX00500090025 PEX00400090025 PEX00600090100 PEX00800160050 PEX00400080050 PEX00600110050 PEX00600110100 PEX02540287050 PEX00600090050 PEX00400080100 PEX01900250050 PEX00500080100 PEX01000140025 PEX00950159050 PEX00400100100 PEX00800140025 PEX00400060100 PEX00630111100 PEX00320063050 PEX00800110050 PEX00630111025 PEX00480079100 PEX00950159100 PEX01900317050 PEX00950127100 PEX00630111050 PEX00600120100 PEX00600080025 PEX00500080025 PEX01200200050 PEX00600080100 PEX00400080025 PEX00480095025 PEX00600100050 PEX01270190025 PEX01270175050 PEX00630095025 PEX00400060050 PEX00600110025 PEX00500110100 PEX00400090100 PEX00600080050 PEX00900150025 PEX01900254050 PEX00400070050 PEX00500090050 PEX01200200025 PEX00630127025 PEX01200160050 PEX00800140050 PEX00300070025 PEX00480079050 PEX00400100025 PEX01270254050 PEX00300070050 PEX00600100025 PEX00480095050 PEX01900286050 PEX00790127100 PEX00600090025 PEX00630095100 PEX00800160025 PEX00500100025 PEX00630095050 PEX00950143100 PEX00630127050 PEX00400070100 PEX01000140050 PEX00800120025 PEX01000160050 PEX00500110050 PEX00800110100 PEX01200160100 PEX01590222050 PEX00600120025 PEX01000160025 PEX00800140100
76517-190PK 125.51 USD InStock
76517-190 76517-198 76517-110 76517-076 76517-230 76517-196 76517-074 76517-194 76517-072 76517-192 76517-070 76517-228 76517-106 76517-226 76517-104 76517-224 76517-102 76517-068 76517-222 76517-188 76517-100 76517-108 76517-080 76517-000 76517-242 76517-088 76517-120 76517-086 76517-240 76517-084 76517-082 76517-118 76517-238 76517-116 76517-236 76517-114 76517-234 76517-112 76517-232 76517-078 76517-090 76517-132 76517-010 76517-098 76517-252 76517-130 76517-250 76517-096 76517-094 76517-092 76517-008 76517-128 76517-006 76517-248 76517-126 76517-004 76517-246 76517-124 76517-244 76517-002 76517-122 76517-022 76517-142 76517-020 76517-140 76517-018 76517-138 76517-016 76517-136 76517-014 76517-134 76517-012 76517-154 76517-032 76517-152 76517-030 76516-986 76517-150 76516-988 76517-028 76517-148 76517-026 76517-146 76517-024 76516-990 76517-144 76517-044 76517-164 76516-992 76517-042 76517-162 76516-994 76517-040 76517-160 76516-996 76516-998 76517-038 76517-158 76517-036 76517-156 76517-034 76517-176 76517-054 76517-174 76517-052 76517-172 76517-050 76517-170 76517-206 76517-204 76517-202 76517-048 76517-168 76517-200 76517-046 76517-166 76517-208 76517-066 76517-220 76517-186 76517-064 76517-184 76517-062 76517-182 76517-060 76517-180 76517-216 76517-214 76517-212 76517-058 76517-178 76517-210 76517-056 76517-218
Imapex™ Peroxide Cured Silicone Tubing, Foxx Life Sciences
Tubing Transfer Tubing
Imapex™ Peroxide Grade silicone tubing is designed for non-critical fluid transfer, peristaltic pump use, and general engineering applications.
  • Manufactured and packaged in dust free environment
  • Odorless and tasteless
  • Enhanced compression set and rensile strength
  • Good flex fatigue resistance enhances peristaltic pump life
Imapex™ is transparent and is made of high-quality peroxide silicone rubber tubing with good physical properties including tensile strength, flexibility, and resiliency. This tubing is manufactured and packaged in dust free environment facility, and has a 3 year shelf life.
Certifications: Meets pharma grade requirement of FDA 21 CFR 177.2600, RoHS certification, Phthalates-/BPA-/Plasticizer-free certification, TSE/BSE free certification.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty