Would you like to log in again?
VWR® Critical Environment Swab, Extra-Large Rectangular Foam Head
Supplier: Avantor
VWR®
SU-131VFCTD
76478-838BX 72.24 USD InStock
76478-838
VWR® Critical Environment Swab, Extra-Large Rectangular Foam Head
Swabs Swabs for Clinical Use
Swab features 100 ppi closed cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam swab head
  • Polypropylene handle
  • Extra-large foam head
Swab head is thermally bonded to the handle without using any adhesive.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty