Would you like to log in again?
Environmental Express® Series 1500 Benchtop Meters, Antylia Scientific
Supplier: Antylia Scientific
35419-41 35419-46 35419-45 35419-40 35419-51 35419-50
76493-628EA 472.6 USD InStock
76493-628 76493-626 76493-624 76493-622 76493-632 76493-630
Environmental Express® Series 1500 Benchtop Meters, Antylia Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Optimized for laboratory measurements such as pH, ORP/mV, conductivity, resistivity, TDS, salinity, and temperature.
  • Large monochrome LCD
  • Integrated electrode holder (up to 2 electrodes) that can be attached to either side of the meter
  • Simple user interface and single parameter display
  • 1000 memory data points
  • Automatic Temperature Compensation (ATC) with temperature calibration
  • Adjustable auto shut-off time (1 to 30 minutes)
  • Auto stable, auto hold, and real time measurement modes with reading stability indicators
  • PC (standard USB) / printer (25 pin serial) connection via 2.5 mm diameter phono jack
  • Buffer recognition
Measurement parameters available vary by model. pH models measure pH and ORP/mV readings. EC models measure conductivity, TDS, salinity, and resistivity. PC models measure pH, ORP/mV, conductivity, TDS, salinity, and resistivity. Users can choose the part number that best fits their needs. In addition to measurement parameters, each model can be bought as a kit or a meter and electrode stand only.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty