Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GSK-2983559

Supplier: BioVision
Synonyms: 2-((4-(benzo[d]thiazol-5-ylamino)-6-(tert-butylsulfonyl)quinazolin-7-yl)oxy)ethyl dihydrogen phosphateGSK29835592-((4-(benzo[d]thiazol-5-ylamino)-6-(tert-butylsulphonyl)quinazolin-7-yl)oxy)ethyl dihydrogen phosphate

AB287132-5MG AB287132-1MG
76472-022EA 557 USD InStock
76472-022 76472-020
GSK-2983559
GSK-2983559
GSK-2983559 is a potent and selective inhibitor of receptor interacting protein-2 (RIP2) kinase.

  • A small-molecule RIP2 kinase inhibitor


Formula: C₂₁H₂₃N₄O₇PS₂
MW: 538,53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30489430
CAS Number: 1579965-12-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty