Would you like to log in again?
Eagle Red Biohazard Waste Bags
Supplier: Eagle Biosciences
Eagle™
1693 1691
76520-562PK 163.46 USD InStock
76520-562 76520-558
Eagle Red Biohazard Waste Bags
Bags Waste Disposal Bags
Holds large amounts of biological waste materials.
  • Red In color
  • Linear low density polyethylene (LLDPE)
  • 1.2 Mil thickness
  • A high level of flexibility.
  • Shear resistance can be attained without leaks
  • Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
For holding or consolidating larger amounts of biological waste or contaminated materials. The 33 gal. capacity bag is designed to fit Eagle 1601 series drums. Once the bag is full and ready for disposal, simply seal the bag in place and close the drum using its lid and lever-lock.
Red color bag is easily identifiable as biohazard bags, drums can be black, yellow, blue or red. Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.
Using Linear Low Density Polyethylene (LLDPE) material at 30.4 µm thickness, a high level of flexibility and shear resistance can be attained without leaks. A tubular construction, single side seal and star seal bottom provide a stable, leak proof bag.
Packaging: Sold in packs of 100.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty