Would you like to log in again?
Adapter for Exhaust Filter for Justrite and CPC Canisters, SCAT Europe
Supplier: S.C.A.T.
107617 107610
76503-208EA 67.17 USD InStock
76503-208 76503-206
Adapter for Exhaust Filter for Justrite and CPC Canisters, SCAT Europe
Fittings and Connectors
Special adapter for connecting a SCAT exhaust air filter with the canisters of the manufacturers Justrite and CPC.
  • Made of PEHD-EC material
  • Fits GL 14 (f) thread on CPC (m) coupling
Compatible for all SCAT exhaust filters sizes small, medium, and large.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty