Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

µ-Plate 96 Well Black, ibidi

Supplier: IBIDI USA MS
89626
MSPP-89626PK 730.77 USD InStock
MSPP-89626
µ-Plate 96 Well Black, ibidi
Microplates Cell Culture Plates
µ-Plate 96 Well Black allows you to perform high resolution microscopy in a standard multi-well format.

The ibidi µ-Plate 96 Well Black is ideal for high resolution imaging through the No. 1.5 polymer coverslip bottom with the highest optical quality while eliminating well-to-well crosstalk in fluorescence microscopy.

Additional specifications:
- Well-to-well distance: 9 mm
- Well clearance: 0.8 mm
- Inner well flatness: ± 5 µm
- Outer well flatness: ± 25 µm
- Depth: 12.9 mm
- Coating area: 2.35 cm² (with 300 µl)
- Growth area: 0.56 cm²

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty