Would you like to log in again?
Brady® BradyJet™ J2000 Continuous Laboratory Tape, Brady
Supplier: Brady
BradyJet™
J20C-4000-7425J
76520-198RL 311.17 USD InStock
76520-198
Brady® BradyJet™ J2000 Continuous Laboratory Tape, Brady
Labels
This B-7425J polypropylene material is ideal for full-color identification and barcode applications where resistance to liquid nitrogen, autoclave conditions, and harsh chemicals is required.
  • Recommended ribbon: J20-CMY
  • Chemical resistant, low temperature labels
  • Permanent acrylic adhesive type
  • Inkjet receptive polypropylene material
Matte white polypropylene with acrylic adhesive is inkjet printable. Perfect for laboratory identification where labels may come in contact with harsh chemicals. Service temperature of −112 to 158 °F (−80 to 70 °C). For use with BradyJet™ J2000 color label printer. An ink cartridge is required to print this material. The recommended ribbon is J20-CMY.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty