Would you like to log in again?
Brady® B33 Series Coated Vinyl Cloth with Repositionable Adhesive Vinyl Labels, Brady
Supplier: Brady
B33
B33-254-498 B33-325-498 B33-326-498 B33-327-498 B33-328-498
76520-320RL 191.75 USD InStock
76520-320 76520-322 76520-324 76520-326 76520-318
Brady® B33 Series Coated Vinyl Cloth with Repositionable Adhesive Vinyl Labels, Brady
Labels
Once applied, the B-498 CleanLift® Series Vinyl Cloth is permanent or repositionable.
  • Recommended ribbon: B30-R6200
  • Repositionable labels
  • Repositionable rubber adhesive type
  • Vinyl cloth material
The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning. Sticks to any clean, dry surface. Specially formulated topcoat for excellent print quality. Semi-gloss finish for a crisp, clean look. A ribbon is required to print this material. The recommended ribbon is B30-R6200.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty