Would you like to log in again?
Brady® B33 Series Vinyl General Identification Labels, Brady
Supplier: Brady
B33
B33-55-439 B33-19-439 B33-18-439 B33-37-439 B33-17-439 B33-76-439 B33-5-439 B33-53-439
76520-288RL 307.14 USD InStock
76520-288 76520-276 76520-278 76520-280 76520-290 76520-282 76520-284 76520-286
Brady® B33 Series Vinyl General Identification Labels, Brady
Labels
Permanent Vinyl (B-439) is a thermal transfer printable material ideal for applications requiring various colors like finished product ID, rating plates, and general purpose ID.
  • Recommended ribbon: B30-R6000
  • Harsh environment label with permanent adhesive
  • Permanent acrylic adhesive type
  • Vinyl material
Designed for use in ambient conditions with limited solvent exposure. Indoor/outdoor vinyl film has an acrylic pressure sensitive adhesive. Service temperature of −94 to 104 °F (−70 to 40 °C). Available in many different colors as a custom option. A ribbon is required to print this material. The recommended ribbon is B30-R6000.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty