Would you like to log in again?
Brady®, B33 Series White Polyester with Permanent Acrylic Adhesive Labels, Brady
Supplier: Brady
B33
B33-343-422 B33-13-422 B33-7-422 B33-2-422 B33-6-422 B33-340-422 B33-51-422 B33-341-422 B33-55-422 B33-53-422 B33-12-422 B33-70-422 B33-37-422 B33-15-422 B33-1-422 B33-3-422
76520-354RL 144.29 USD InStock
76520-354 76520-376 76520-364 76520-378 76520-356 76520-366 76520-358 76520-368 76520-370 76520-380 76520-372 76520-360 76520-382 76520-352 76520-374 76520-362
Brady®, B33 Series White Polyester with Permanent Acrylic Adhesive Labels, Brady
Labels
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Recommended ribbon: B30-R6000
  • Heat resistant label with permanent adhesive
  • Permanent acrylic adhesive type
  • Polyester material
  • UL approved, CSA accepted
Aggressive adhesive is good for powder-coated and textured flat surfaces including low surface energy plastics. Designed to withstand exposure to numerous solvents. Service temperature of −40 to 212 °F (−40 to 100 °C). A ribbon is required to print this material. The recommended ribbon is B30-R6000.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty