Would you like to log in again?
Brady® R6200 Series Printer Ribbon, Brady
Supplier: Brady
M61-R6210
76520-188RL 45.25 USD InStock
76520-188
Brady® R6200 Series Printer Ribbon, Brady
Labels
BMP®61 Series ribbon cartridges allow users to change ribbons in seconds.
  • For use with BMP®61 label printer and BradyPrinter M611 mobile label printer
One-color ribbon for printing a single color onto a label. R6200 Series Ribbons offer superior scratch, smear, heat, and chemical resistance when printed on the appropriate label material from Brady®. Recommended for printing on glossy, smooth, or semi-matte label stock, such as B-498 Repositionable Vinyl Cloth and B-486 Metallized Polyester. Automatic label formatting with smart cell technology and easy loading.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty