Would you like to log in again?
Air Valves for SafetyCaps Generation 2.0
Supplier: S.C.A.T.
397008 397100 317010 397050 397010
76503-974PK 416.82 USD InStock
76503-974 76948-972 76503-852 76503-976 76948-970
Air Valves for SafetyCaps Generation 2.0
Valves Air Regulation Valves
These air valves can be used for analytical and preperative applications.
  • PTFE filter for clean solvents
  • Valve membrane blocks solvent vapours, opens at underpressure
  • Thread size UNF 1/4" 28G
They include a luer lock adapter which is useful for many applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty