Would you like to log in again?
NIGHTSEA Fluorescence Adapter for Leica, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
SFA-DVM6-RB SFA-DVM6-UV SFADVM6-FSGR SFADVM6LFSCY SFA-DVM6-VI SFADVM6LFSUV SFADVM6LFSRB SFADVM6LFSVI SFA-DVM6-CY SFA-DVM6-GR
76499-848EA 2392.17 USD InStock
76499-848 76499-844 76499-834 76499-846 76499-836 76499-840 76499-852 76499-842 76499-832 76499-850
NIGHTSEA Fluorescence Adapter for Leica, Electron Microscopy Sciences
Lights
NIGHTSEA SFA system can add versatile fluorescence imaging capability to the Leica DVM6 microscope.
  • High intensity LED light sources available in five excitation wavelength ranges
  • Emission barrier filters attach to the DVM6 objectives easily and non-invasively
DVM6 fluorescence adapter kit that includes the most useful excitation wavelengths for varied industry applications.
Each kit includes three excitation light heads (ultraviolet (UV), violet (VI), and royal blue (RB)), three barrier filters and filter shields paired to the excitation light heads, gooseneck lamp base with intensity control, universal power supply (120/240 VAC, 50/60 Hz), with international plug set and a packing/carry case with custom cut form.
Ordering information: Additional wavelength sets can be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty