Would you like to log in again?
NIGHTSEA Mobile Extension Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
SFA-MEK-RB-GO SFA-MEK-UV SFA-MEK-GR SFA-MEK-CY SFA-MEK-RB
76499-866EA 426.33 USD InStock
76499-866 76499-868 76499-862 76499-864 76499-860
NIGHTSEA Mobile Extension Kit, Electron Microscopy Sciences
Lights
Mobile Extension kit provides added versatility to the NIGHTSEA SFA Adapter system.
This Stereo Microscope Fluorescence Adapter enables users to utilize the light head as a handheld light source.
Each kit includes a rubber-coated hand grip with wrist lanyard and custom enclosure that holds an SFA Light Head securely. The slots on the side of the enclosure provide for cooling and allow viewing the color code of an installed light head. Lightweight extension cable and barrier filter glasses included.
The SFA-MEK is the only requirement when using the Mobile Extension Kit with an existing SFA system. To add additional excitation/emission combinations to use only for handheld operation (not for a microscope), a Light Head plus a pair of Barrier Filter Glasses for each color is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty