Would you like to log in again?
µ-Slide 2-Well Co-Culture Slide, Ibidi
Supplier: IBIDI USA MS
81806
MSPP-81806EA 369.23 USD InStock
MSPP-81806
µ-Slide 2-Well Co-Culture Slide, Ibidi
Slides Chamber Slides
A µ-Slide, chambered coverslip suitable for co-cultivation assays in combination with microscopy.
  • Cells share soluble factors but grow separately
  • Excellent optical quality suitable for fluorescence microscopy
  • Coating area: 0.55 cm² (per minor well)
  • Well dimensions: 6.1W×6.8L×1.3H mm³ (minor wells), 21.5W×23.6L×6.8H mm³ (major wells)
Possesses low evaporation properties and is suitable for long-term assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty