Would you like to log in again?
Bond Elut C8, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Bond Elut™
12256002 12113025 12256002 12113025
AG12256002PK 169 USD InStock
CAAG12256002 CAAG12113025 AG12256002 AG12113025
Bond Elut C8, Agilent Technologies
Chromatography Columns
Bond Elut C8 is a good alternative when analytes are too strongly retained on C18 phases.
  • Excellent for strongly retained analytes
  • Polar interactions are not significant
  • Less retentive than C18
Bond Elut C8 is very similar in property to C18, but is not as retentive for nonpolar compounds due to its shorter hydrocarbon chain and therefore reduced carbon loading. C8 is an excellent replacement for C18 when analytes are too strongly retained for effective elution. The potential for polar interactions is higher than for C18 because there is less coverage of the silica surface.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty