Would you like to log in again?
Bond Elut ENV, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12255012 12255012
AG12255012PK 277 USD InStock
AG12255012 CAAG12255012
Bond Elut ENV, Agilent Technologies
Chromatography Columns
Bond Elut ENV is a large particle size phase that allows fast extraction of highly polar compounds.
  • Modified styrene-divinylbenzene polymer
  • High surface area and capacity for polar analytes
  • Large particle size allows high flowthrough
  • Efficient elution characteristics
Bond Elut ENV, a PS-DVB polymer, is designed for the extraction of polar organic residues. It contains 125 µm spherical particles with a high degree of cross-linking, advantageous for high-volume, fast-flowthrough applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty