Would you like to log in again?
Bond Elut NH₂, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
12102089 14102076 14256012 14113040 14256012 14113040 12113040 12102106 12102106 12102089 14102076 12113040
AG12102106PK 223 USD InStock
AG12102106 CAAG14256012 AG14102076 AG14256012 CAAG12102106 CAAG14102076 AG14113040 CAAG14113040 CAAG12113040 CAAG12102089 AG12113040 AG12102089
Bond Elut NH₂, Agilent Technologies
Chromatography Columns
Bond Elut NH2 is a weak anion exchanger and a good choice for retention of very strong anions.
  • Normal phase or anion-exchange sorbent
  • Weaker anion exchange than SAX
  • Amenable to separating structural isomers
Bond Elut NH2 is a weaker anion exchanger than sorbents such as SAX and is therefore a better choice for retention of very strong anions. Similar to Bond Elut Diol and SI sorbents, Bond Elut NH2 is excellent for the separation of structural isomers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty