Would you like to log in again?
Membrane Filters, Type 131, Sartorius
Supplier: Sartorius
1310150AHN 1310150AHN
76500-650PK 265.03 USD InStock
CA76517-688 76500-650
Membrane Filters, Type 131, Sartorius
Filters Membrane Filters
Non sterile membrane filters for colony counting, particle testing, microscopy and sterility testing. The membrane filters are made of cellulose nitrate (cellulose ester).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty