Would you like to log in again?
Spray Nine Heavy-Duty Cleaner/Degreaser/Disinfectant, Citrus Scent, Essendant
Supplier: Essendant
ITW268014CT
76500-326CT 138.05 USD InStock
76500-326
Spray Nine Heavy-Duty Cleaner/Degreaser/Disinfectant, Citrus Scent, Essendant
Cleaning Agents Disinfectants
A professional-grade cleaner and disinfectant that removes all types of dirt, grease, grime, and stains.
  • Professional-grade cleaner, degreaser, and disinfectant in one
  • Disinfects against viruses in 30 seconds
  • Disinfect against bacteria in 45 seconds
  • Sanitizes in 10 seconds
  • Kills fungus in three minutes
  • Deodorizes
  • Controls mold and mildew
  • Removes stains
One product with nine uses, this unique formula cleans, degreases, disinfects against viruses, disinfects against bacteria, sanitizes, kills fungus, deodorizes, controls mold and mildew, and removes stains. Suitable for use in bathrooms, breakrooms, kitchens, offices, manufacturing plants, medical offices and exam rooms, hospitals/healthcare facilities, and veterinary facilities. Chemical compound: N-Alkyl Dimethyl Benzyl Ammonium Chloride; N-Alkyl Dimethyl Ethybenzyl Ammonium Chloride; Ethoxylated Alcohols; Dipropylene Glycol Mononbutyl Ether.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty