Would you like to log in again?
Goo Gone Pro-Power Cleaner, Citrus Scent, Essendant
Supplier: Essendant
Goo Gone
WMN2085
76500-332EA 54.96 USD InStock
76500-332
Goo Gone Pro-Power Cleaner, Citrus Scent, Essendant
Cleaning Agents Detergents and Surface Cleansers
Pro Power formula is enhanced to tackle the toughest jobs.
  • Surface-safe and fast-working remover
Pro Power formula can be used to remove silicone caulk, contractor's adhesive, grease, tar, tape, tape residue, varnish, wax, and glue. Designed for use on hard non-porous surfaces and soft fabric surfaces like carpet, clothing, and upholstery. Chemical compound: Hydrotreated light distillates; Tripropylene glycol methyl ether.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty