Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boardwalk® Spring Grip Metal Head Wooden Mop Handle, Essendant

Supplier: Essendant
Boardwalk®
BWK609
76500-342EA 39.57 USD InStock
76500-342
Boardwalk® Spring Grip Metal Head Wooden Mop Handle, Essendant
Mops
Rust-resistant 0.25" wire securely locks most mop heads in place.

  • Handle diameter: 1.13"
  • Handle length: 60"

Double-lacquered wooden mop handle in a natural finish. Designed for use with most metal mop heads.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty