Would you like to log in again?
Rubbermaid® Super Stitch Looped-End Wet Mop Head, Cotton/Synthetic, Essendant
Supplier: Essendant
Rubbermaid®
RCPD252WHI
76500-388CT 73.53 USD InStock
76500-388
Rubbermaid® Super Stitch Looped-End Wet Mop Head, Cotton/Synthetic, Essendant
Mops
The Super Stitch Blend Mop has a blend of 4-ply cotton and synthetic fibers for higher absorbency and more launderings than cotton mops, and has looped ends and a yarn tailband to reduce tangling and fraying for improved performance.
  • Balanced blend of cotton and synthetic yarns for absorbency and strength
  • Looped ends and yarn tailband mop for greater floor coverage
  • Suitable for use in commercial and industrial applications
The synthetic mesh headband resists moisture retention so the mop head can dry between uses. Green cleaning equipment is recognized in the industry as color-coded for use in food service areas as part of a zone isolation system to help minimize cross-contamination. These mop heads are suitable for use in commercial and industrial applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty