Would you like to log in again?
Clorox® Ultra Clean Toilet Tablets, Essendant
Supplier: Essendant
Clorox®
CLO30024CT
76500-090CT 82.32 USD InStock
76500-090
Clorox® Ultra Clean Toilet Tablets, Essendant
Cleaning Agents Disinfectants
Clorox® Ultra Clean Toilet Tablets kill 99.9% of bacteria and deodorizes the toilet with the power of Clorox™ bleach.
  • Make cleaning easy and hassle free with this sanitizing automatic toilet cleaner
  • Flushable toilet cleaning tablets continuously clean with bleach and deodorize the bathroom
  • Prevent tough stains, including hard water and mineral stains, with the power of Clorox® cleaner
  • Bleach in every flush helps extend the time between deep cleans
  • Will not harm septic tanks
  • Chemical compound: 1-bromo-3-chloro-5,5-dimethylhydantoin 51.0%
This toilet bowl cleaning tablet effectively cleans and prevents stains with each flush, eliminating the need to scrub every time. Each tablet delivers a long-lasting bleach clean. Simply drop a tablet into the toilet tank to continuously sanitize, deodorize, and clean the toilet with every flush. The dissolvable cleaning tablets help prevent tough stains and removes hard water, mineral deposits, and limescale stains. Ideal for use in restaurants, cafeterias, offices, hotels, daycares, and other commercial facilities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty