Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ProLysol Disinfectant Toilet Bowl Cleaner, Essendant
Supplier: Essendant
RAC74278CT
76500-134CT 90.82 USD InStock
76500-134
ProLysol Disinfectant Toilet Bowl Cleaner, Essendant
Cleaning Agents Disinfectants
Kills Staph, Pseudomonas, E. coli, HIV-1 (AIDS virus) and other harmful germs on hard, non-porous surfaces when used as directed.
  • Cleans, disinfects, and deodorizes toilets and urinals
  • Kills 99.9% of viruses and bacteria when used as directed
  • Angle-neck bottle targets hard-to-reach areas
  • Will not harm plumbing or septic systems
The thick 9.5% hydrochloric acid formula coats, cleans, disinfects, and deodorizes the entire toilet bowl by dissolving stubborn stains on contact. The angle-neck bottle targets hard-to-reach areas. Formula will not harm plumbing or septic systems. Use on toilets and urinals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty