Would you like to log in again?
Dymon Medaphene Plus Disinfectant Spray, Essendant
Supplier: Essendant
ITW35720
76500-138CT 191.99 USD InStock
76500-138
Dymon Medaphene Plus Disinfectant Spray, Essendant
Cleaning Agents Disinfectants
Bactericidal, tuberculocidal, fungicidal, and virucidal spray effectively eliminates a wide range of disease-causing organisms.
  • Deodorizes as it disinfects
  • Easy to use
  • Chemical compound: Ethyl Alcohol; N-Butane; O-Phenylphenol; Propane
While it disinfects, this aerosol spray deodorizes areas contaminated by most organic sources of malodors. Use on countertops, door handles, faucets, hard surfaces, railings, showers, sinks, telephones, toilets, trash cans, and urinals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty